CD39 Monoclonal Antibody (24DMS1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD39 Monoclonal Antibody (24DMS1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD39 Monoclonal Antibody (24DMS1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0391-80, 46-0391-82)