CD40 Monoclonal Antibody (5C3), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD40 Monoclonal Antibody (5C3), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD40 Monoclonal Antibody (5C3), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0409-42, 46-0409-41)