CD41a Monoclonal Antibody (HIP8), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD41a Monoclonal Antibody (HIP8), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD41a Monoclonal Antibody (HIP8), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0419-41, 46-0419-42)