CD42d Monoclonal Antibody (1C2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD42d Monoclonal Antibody (1C2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD42d Monoclonal Antibody (1C2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0421-82, 46-0421-80)