CD45R (B220) Monoclonal Antibody (RA3-6B2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD45R (B220) Monoclonal Antibody (RA3-6B2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD45R (B220) Monoclonal Antibody (RA3-6B2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0452-80, 46-0452-82)