CD45RO Monoclonal Antibody (UCHL1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD45RO Monoclonal Antibody (UCHL1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD45RO Monoclonal Antibody (UCHL1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0457-42, 46-0457-41)