MHC Class II Monoclonal Antibody (OX17), PerCP-eFluor™ 710, eBioscience™ - FAQs
MHC Class II Monoclonal Antibody (OX17), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for MHC Class II Monoclonal Antibody (OX17), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0463-82)