CD45RA Monoclonal Antibody (GRT22), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD45RA Monoclonal Antibody (GRT22), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD45RA Monoclonal Antibody (GRT22), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0468-42)