CD46 Monoclonal Antibody (8E2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD46 Monoclonal Antibody (8E2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD46 Monoclonal Antibody (8E2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0469-42, 46-0469-41)