CD47 Monoclonal Antibody (B6H12), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD47 Monoclonal Antibody (B6H12), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD47 Monoclonal Antibody (B6H12), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0479-42)