CD48 Monoclonal Antibody (HM48-1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD48 Monoclonal Antibody (HM48-1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD48 Monoclonal Antibody (HM48-1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0481-82, 46-0481-80)