CD64 Monoclonal Antibody (X54-5/7.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD64 Monoclonal Antibody (X54-5/7.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD64 Monoclonal Antibody (X54-5/7.1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0641-80, 46-0641-82)