CD64 Monoclonal Antibody (10.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD64 Monoclonal Antibody (10.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD64 Monoclonal Antibody (10.1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0649-42, 46-0649-41)