CD69 Monoclonal Antibody (FN50), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD69 Monoclonal Antibody (FN50), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD69 Monoclonal Antibody (FN50), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0699-41, 46-0699-42)