CD70 Monoclonal Antibody (FR70), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD70 Monoclonal Antibody (FR70), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD70 Monoclonal Antibody (FR70), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0701-80, 46-0701-82)