CD73 Monoclonal Antibody (AD2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD73 Monoclonal Antibody (AD2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD73 Monoclonal Antibody (AD2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0739-41, 46-0739-42)