CD81 Monoclonal Antibody (1D6-CD81), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD81 Monoclonal Antibody (1D6-CD81), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD81 Monoclonal Antibody (1D6-CD81), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0819-42, 46-0819-41)