CD83 Monoclonal Antibody (HB15e), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD83 Monoclonal Antibody (HB15e), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD83 Monoclonal Antibody (HB15e), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0839-42)