CD90.2 (Thy-1.2) Monoclonal Antibody (30-H12), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD90.2 (Thy-1.2) Monoclonal Antibody (30-H12), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD90.2 (Thy-1.2) Monoclonal Antibody (30-H12), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0903-80, 46-0903-82)