CD91 Monoclonal Antibody (A2MR-a2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD91 Monoclonal Antibody (A2MR-a2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD91 Monoclonal Antibody (A2MR-a2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0919-42, 46-0919-41)