CD95 (APO-1/Fas) Monoclonal Antibody (DX2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD95 (APO-1/Fas) Monoclonal Antibody (DX2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD95 (APO-1/Fas) Monoclonal Antibody (DX2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-0959-42)