VISTA Monoclonal Antibody (MIH64), PerCP-eFluor™ 710, eBioscience™ - FAQs
VISTA Monoclonal Antibody (MIH64), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for VISTA Monoclonal Antibody (MIH64), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1083-82, 46-1083-80)