CD122 Monoclonal Antibody (TU27), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD122 Monoclonal Antibody (TU27), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD122 Monoclonal Antibody (TU27), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1228-42, 46-1228-41)