CD126 Monoclonal Antibody (47.7G7.1F2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD126 Monoclonal Antibody (47.7G7.1F2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD126 Monoclonal Antibody (47.7G7.1F2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1269-42, 46-1269-41)