CD135 (Flt3) Monoclonal Antibody (A2F10), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD135 (Flt3) Monoclonal Antibody (A2F10), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD135 (Flt3) Monoclonal Antibody (A2F10), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1351-80, 46-1351-82)