CD144 (VE-cadherin) Monoclonal Antibody (eBioBV13 (BV13)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD144 (VE-cadherin) Monoclonal Antibody (eBioBV13 (BV13)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD144 (VE-cadherin) Monoclonal Antibody (eBioBV13 (BV13)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1441-80, 46-1441-82)