CD150 Monoclonal Antibody (mShad150), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD150 Monoclonal Antibody (mShad150), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD150 Monoclonal Antibody (mShad150), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1502-82)