CD154 (CD40 Ligand) Monoclonal Antibody (24-31), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD154 (CD40 Ligand) Monoclonal Antibody (24-31), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD154 (CD40 Ligand) Monoclonal Antibody (24-31), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1548-42, 46-1548-41)