CD155 Monoclonal Antibody (2H7CD155), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD155 Monoclonal Antibody (2H7CD155), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD155 Monoclonal Antibody (2H7CD155), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1550-41, 46-1550-42)