CD161 Monoclonal Antibody (10/78), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD161 Monoclonal Antibody (10/78), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1610-82, 46-1610-80)