CD163 Monoclonal Antibody (TNKUPJ), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD163 Monoclonal Antibody (TNKUPJ), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD163 Monoclonal Antibody (TNKUPJ), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1631-82, 46-1631-80)