CD182 (CXCR2) Monoclonal Antibody (eBio5E8-C7-F10 (5E8-C7-F10)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD182 (CXCR2) Monoclonal Antibody (eBio5E8-C7-F10 (5E8-C7-F10)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD182 (CXCR2) Monoclonal Antibody (eBio5E8-C7-F10 (5E8-C7-F10)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-1829-42, 46-1829-41)