CD201 (EPCR) Monoclonal Antibody (eBio1560 (1560)), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD201 (EPCR) Monoclonal Antibody (eBio1560 (1560)), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD201 (EPCR) Monoclonal Antibody (eBio1560 (1560)), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-2012-80)