CD205 Monoclonal Antibody (205yekta), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD205 Monoclonal Antibody (205yekta), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD205 Monoclonal Antibody (205yekta), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-2051-80, 46-2051-82)