CD206 (MMR) Monoclonal Antibody (19.2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD206 (MMR) Monoclonal Antibody (19.2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD206 (MMR) Monoclonal Antibody (19.2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-2069-42, 46-2069-41)