CD319 (CRACC) Monoclonal Antibody (162), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD319 (CRACC) Monoclonal Antibody (162), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD319 (CRACC) Monoclonal Antibody (162), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-2229-41, 46-2229-42)