CD243 (ABCB1) Monoclonal Antibody (UIC2), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD243 (ABCB1) Monoclonal Antibody (UIC2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD243 (ABCB1) Monoclonal Antibody (UIC2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-2439-42)