CD244.1 Monoclonal Antibody (C9.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD244.1 Monoclonal Antibody (C9.1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD244.1 Monoclonal Antibody (C9.1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-2440-82, 46-2440-80)