CD276 (B7-H3) Monoclonal Antibody (7-517), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD276 (B7-H3) Monoclonal Antibody (7-517), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD276 (B7-H3) Monoclonal Antibody (7-517), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-2769-41, 46-2769-42)