CD304 (Neuropilin-1) Monoclonal Antibody (TNKUSOHA), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD304 (Neuropilin-1) Monoclonal Antibody (TNKUSOHA), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD304 (Neuropilin-1) Monoclonal Antibody (TNKUSOHA), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-3049-41, 46-3049-42)