CD307d (FcRL4) Monoclonal Antibody (413D12), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD307d (FcRL4) Monoclonal Antibody (413D12), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD307d (FcRL4) Monoclonal Antibody (413D12), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-3079-42, 46-3079-41)