CD335 (NKp46) Monoclonal Antibody (9E2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD335 (NKp46) Monoclonal Antibody (9E2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-3359-41, 46-3359-42)