CD370 (Clec9A) Monoclonal Antibody (9A11), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD370 (Clec9A) Monoclonal Antibody (9A11), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD370 (Clec9A) Monoclonal Antibody (9A11), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-3709-41, 46-3709-42)