Rat anti-Mouse IgG1 Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs
Rat anti-Mouse IgG1 Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Rat anti-Mouse IgG1 Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4015-82)