Rat anti-Mouse IgG2a Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs
Rat anti-Mouse IgG2a Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Rat anti-Mouse IgG2a Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4210-82, 46-4210-80)