Rat IgG1 kappa Isotype Control (eBRG1), PerCP-eFluor™ 710, eBioscience™ - FAQs
Rat IgG1 kappa Isotype Control (eBRG1), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Rat IgG1 kappa Isotype Control (eBRG1), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4301-80)