Rat IgG2a kappa Isotype Control (eBR2a), PerCP-eFluor™ 710, eBioscience™ - FAQs
Rat IgG2a kappa Isotype Control (eBR2a), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Rat IgG2a kappa Isotype Control (eBR2a), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4321-82, 46-4321-80)