Rat IgM Isotype Control (eBRM), PerCP-eFluor™ 710, eBioscience™ - FAQs
Rat IgM Isotype Control (eBRM), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Rat IgM Isotype Control (eBRM), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4341-80)