Mouse anti-Rat IgG2a Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Mouse anti-Rat IgG2a Secondary Antibody, PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4817-82, 46-4817-80)