EOMES Monoclonal Antibody (Dan11mag), PerCP-eFluor™ 710, eBioscience™ - FAQs
EOMES Monoclonal Antibody (Dan11mag), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for EOMES Monoclonal Antibody (Dan11mag), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-4875-82, 46-4875-80)